tert-Butyl-4-(2-(trifluoromethyl)-1,6-naphthyridin-5-ylamino)piperidine-1-carboxylate

Supplier: Matrix Scientific

032302-250MG
101833-422EA 315.38 USD
101833-422
tert-Butyl-4-(2-(trifluoromethyl)-1,6-naphthyridin-5-ylamino)piperidine-1-carboxylate
tert-Butyl-4-(2-(trifluoromethyl)-1,6-naphthyridin-5-ylamino)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₉H₂₃F₃N₄O₂
MW: 396.42 g/mol
MDL Number: MFCD10688282
CAS Number: 1040682-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR