5-Cyclopropyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one

Supplier: Matrix Scientific

032299-250MG
101833-416EA 218.46 USD
101833-416
5-Cyclopropyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one
5-Cyclopropyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one

Hazard: Irritant

Formula: C₇H₈N₂OS
MDL Number: MFCD10688279
CAS Number: 59698-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR