3,5-Bis(methylsulfonyl)benzylamine hydrochloride

Supplier: Matrix Scientific

032289-500MG
101833-400EA 266.15 USD
101833-400
3,5-Bis(methylsulfonyl)benzylamine hydrochloride
3,5-Bis(methylsulfonyl)benzylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClNO₄S₂
MW: 299.8 g/mol
MDL Number: MFCD10688274
CAS Number: 1171787-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR