tert-Butyl-4-(aminomethyl)phenyl(methyl)carbamate

Supplier: Matrix Scientific

032288-250MG
101833-398EA 250.77 USD
101833-398
tert-Butyl-4-(aminomethyl)phenyl(methyl)carbamate
tert-Butyl-4-(aminomethyl)phenyl(methyl)carbamate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MW: 236.32 g/mol
MDL Number: MFCD09953964
CAS Number: 191871-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR