4-(Imidazol-1-yl)benzylamine

Supplier: Matrix Scientific

032281-500MG
101833-386EA 164.62 USD
101833-386
4-(Imidazol-1-yl)benzylamine
4-(Imidazol-1-yl)benzylamine

Hazard: Irritant

Formula: C₁₀H₁₁N₃
MW: 173.22 g/mol
MDL Number: MFCD06213154
CAS Number: 65113-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR