[R,S]-tert-Butyl-3-amino-2-methylpropionate

Supplier: Matrix Scientific

032278-250MG
101833-380EA 287.69 USD
101833-380
[R,S]-tert-Butyl-3-amino-2-methylpropionate
[R,S]-tert-Butyl-3-amino-2-methylpropionate

Hazard: Irritant

Formula: C₈H₁₇NO₂
MW: 159.23 g/mol
MDL Number: MFCD10688272
CAS Number: 120378-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR