Imidazo[1,2-a]pyridine-2-carbaldehyde
Supplier: Matrix Scientific
Synonyms:
Imidazo[1,2-a]pyridine-2-carboxaldehyde
032277-500MG
101833-378EA
340
USD
101833-378
Imidazo[1,2-a]pyridine-2-carbaldehyde
Imidazo[1,2-a]pyridine-2-carbaldehyde
Hazard: Irritant
Formula:
C₈H₆N₂O MW: 146.1460 g/mol |
MDL Number:
MFCD06739247 CAS Number: 118000-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...