3-(Tetrahydro-2-furanylmethoxy)-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]aniline

Supplier: Matrix Scientific

032266-500MG
101833-356EA 306.15 USD
101833-356
3-(Tetrahydro-2-furanylmethoxy)-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]aniline
3-(Tetrahydro-2-furanylmethoxy)-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₃H₂₉NO₄
MDL Number: MFCD10688266
CAS Number: 1040682-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR