N-[4-(Phenethyloxy)benzyl]-3-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

032260-500MG
101833-344EA 306.15 USD
101833-344
N-[4-(Phenethyloxy)benzyl]-3-(tetrahydro-2-furanylmethoxy)aniline
N-[4-(Phenethyloxy)benzyl]-3-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₂₆H₂₉NO₃
MDL Number: MFCD10688260
CAS Number: 1040681-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR