N-[2-(4-Isopropylphenoxy)propyl]-3-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

032259-500MG
101833-342EA 306.15 USD
101833-342
N-[2-(4-Isopropylphenoxy)propyl]-3-(tetrahydro-2-furanylmethoxy)aniline
N-[2-(4-Isopropylphenoxy)propyl]-3-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₂₃H₃₁NO₃
MDL Number: MFCD10688259
CAS Number: 1040681-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR