N-[2-(Hexyloxy)benzyl]-3-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

032257-500MG
101833-338EA 306.15 USD
101833-338
N-[2-(Hexyloxy)benzyl]-3-(tetrahydro-2-furanylmethoxy)aniline
N-[2-(Hexyloxy)benzyl]-3-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₂₄H₃₃NO₃
MDL Number: MFCD10688257
CAS Number: 1040681-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR