N-Butyl-3-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

032256-500MG
101833-336EA 306.15 USD
101833-336
N-Butyl-3-(tetrahydro-2-furanylmethoxy)aniline
N-Butyl-3-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₁₅H₂₃NO₂
MDL Number: MFCD10688256
CAS Number: 1040681-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR