N-(4-Fluorobenzyl)-3-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

032254-500MG
101833-332EA 306.15 USD
101833-332
N-(4-Fluorobenzyl)-3-(tetrahydro-2-furanylmethoxy)aniline
N-(4-Fluorobenzyl)-3-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₁₈H₂₀FNO₂
MDL Number: MFCD10688254
CAS Number: 1040681-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR