3-Methoxy-N-(2-methylphenethyl)-1-propanamine

Supplier: Matrix Scientific

032252-500MG
101833-328EA 306.15 USD
101833-328
3-Methoxy-N-(2-methylphenethyl)-1-propanamine
3-Methoxy-N-(2-methylphenethyl)-1-propanamine

Hazard: Irritant

Formula: C₁₃H₂₁NO
MDL Number: MFCD10688252
CAS Number: 1040681-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR