N-(3-Methoxypropyl)-1-hexanamine

Supplier: Matrix Scientific

032248-500MG
101833-320EA 306.15 USD
101833-320
N-(3-Methoxypropyl)-1-hexanamine
N-(3-Methoxypropyl)-1-hexanamine

Hazard: Irritant

Formula: C₁₀H₂₃NO
MDL Number: MFCD10688248
CAS Number: 111106-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR