N-[2-(2-Chlorophenoxy)ethyl]-3-methoxy-1-propanamine

Supplier: Matrix Scientific

032246-500MG
101833-316EA 306.15 USD
101833-316
N-[2-(2-Chlorophenoxy)ethyl]-3-methoxy-1-propanamine
N-[2-(2-Chlorophenoxy)ethyl]-3-methoxy-1-propanamine

Hazard: Irritant

Formula: C₁₂H₁₈ClNO₂
MDL Number: MFCD10688246
CAS Number: 1040680-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR