N-[4-(2,4-Dichlorophenoxy)butyl]-N-(3-methoxypropyl)amine

Supplier: Matrix Scientific

032244-500MG
101833-312EA 306.15 USD
101833-312
N-[4-(2,4-Dichlorophenoxy)butyl]-N-(3-methoxypropyl)amine
N-[4-(2,4-Dichlorophenoxy)butyl]-N-(3-methoxypropyl)amine

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO₂
MDL Number: MFCD10688244
CAS Number: 1040680-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR