N1-[2-(3,4-Dimethylphenoxy)ethyl]-N4,N4-diethyl-1,4-benzenediamine

Supplier: Matrix Scientific

032240-500MG
101833-304EA 306.15 USD
101833-304
N1-[2-(3,4-Dimethylphenoxy)ethyl]-N4,N4-diethyl-1,4-benzenediamine
N1-[2-(3,4-Dimethylphenoxy)ethyl]-N4,N4-diethyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₂₀H₂₈N₂O
MDL Number: MFCD10688240
CAS Number: 1040680-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR