3-(Heptyloxy)-N-[2-(2-phenoxyethoxy)benzyl]aniline

Supplier: Matrix Scientific

032238-500MG
101833-300EA 306.15 USD
101833-300
3-(Heptyloxy)-N-[2-(2-phenoxyethoxy)benzyl]aniline
3-(Heptyloxy)-N-[2-(2-phenoxyethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₈H₃₅NO₃
MDL Number: MFCD10688238
CAS Number: 1040680-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR