3-(Heptyloxy)-N-(2-propoxybenzyl)aniline
Supplier: Matrix Scientific
032236-500MG
101833-296EA
306.15
USD
101833-296
3-(Heptyloxy)-N-(2-propoxybenzyl)aniline
3-(Heptyloxy)-N-(2-propoxybenzyl)aniline
Hazard: Irritant
Formula:
C₂₃H₃₃NO₂ |
MDL Number:
MFCD10688236 CAS Number: 1040680-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...