3-(Heptyloxy)-N-[2-(2-methylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

032233-500MG
101833-290EA 306.15 USD
101833-290
3-(Heptyloxy)-N-[2-(2-methylphenoxy)ethyl]aniline
3-(Heptyloxy)-N-[2-(2-methylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₂₂H₃₁NO₂
MDL Number: MFCD10688233
CAS Number: 1040680-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR