N-[4-(2-Cyclohexylethoxy)phenyl]-N-(4-isopropylbenzyl)amine

Supplier: Matrix Scientific

032230-500MG
101833-284EA 306.15 USD
101833-284
N-[4-(2-Cyclohexylethoxy)phenyl]-N-(4-isopropylbenzyl)amine
N-[4-(2-Cyclohexylethoxy)phenyl]-N-(4-isopropylbenzyl)amine

Hazard: Irritant

Formula: C₂₄H₃₃NO
MDL Number: MFCD10688230
CAS Number: 1040679-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR