N-[2-(2-Chlorophenoxy)propyl]-4-(2-cyclohexylethoxy)aniline

Supplier: Matrix Scientific

032225-500MG
101833-274EA 306.15 USD
101833-274
N-[2-(2-Chlorophenoxy)propyl]-4-(2-cyclohexylethoxy)aniline
N-[2-(2-Chlorophenoxy)propyl]-4-(2-cyclohexylethoxy)aniline

Hazard: Irritant

Formula: C₂₃H₃₀ClNO₂
MDL Number: MFCD10688225
CAS Number: 1040693-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR