4-(2-Cyclohexylethoxy)-N-neopentylaniline

Supplier: Matrix Scientific

032224-500MG
101833-272EA 423.08 USD
101833-272
4-(2-Cyclohexylethoxy)-N-neopentylaniline
4-(2-Cyclohexylethoxy)-N-neopentylaniline

Hazard: Irritant

Formula: C₁₉H₃₁NO
MDL Number: MFCD10688224
CAS Number: 1040693-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR