N-[4-(2-Cyclohexylethoxy)phenyl]-N-(3-phenylpropyl)amine

Supplier: Matrix Scientific

032220-500MG
101833-264EA 306.15 USD
101833-264
N-[4-(2-Cyclohexylethoxy)phenyl]-N-(3-phenylpropyl)amine
N-[4-(2-Cyclohexylethoxy)phenyl]-N-(3-phenylpropyl)amine

Hazard: Irritant

Formula: C₂₃H₃₁NO
MDL Number: MFCD10688220
CAS Number: 1040693-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR