N-(4-Chlorobenzyl)-4-(2-cyclohexylethoxy)aniline

Supplier: Matrix Scientific

032219-500MG
101833-262EA 306.15 USD
101833-262
N-(4-Chlorobenzyl)-4-(2-cyclohexylethoxy)aniline
N-(4-Chlorobenzyl)-4-(2-cyclohexylethoxy)aniline

Hazard: Irritant

Formula: C₂₁H₂₆ClNO
MDL Number: MFCD10688219
CAS Number: 1040693-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR