N-(2-Chlorobenzyl)-4-(2-cyclohexylethoxy)aniline

Supplier: Matrix Scientific

032218-500MG
101833-260EA 306.15 USD
101833-260
N-(2-Chlorobenzyl)-4-(2-cyclohexylethoxy)aniline
N-(2-Chlorobenzyl)-4-(2-cyclohexylethoxy)aniline

Hazard: Irritant

Formula: C₂₁H₂₆ClNO
MDL Number: MFCD10688218
CAS Number: 1040693-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR