N1,N1-Diethyl-N4-(2-phenoxyethyl)-1,4-benzenediamine

Supplier: Matrix Scientific

032204-500MG
101833-232EA 306.15 USD
101833-232
N1,N1-Diethyl-N4-(2-phenoxyethyl)-1,4-benzenediamine
N1,N1-Diethyl-N4-(2-phenoxyethyl)-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₈H₂₄N₂O
MDL Number: MFCD03210893
CAS Number: 356532-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR