N1,N1-Diethyl-N4-(3-phenylpropyl)-1,4-benzenediamine

Supplier: Matrix Scientific

032200-500MG
101833-224EA 306.15 USD
101833-224
N1,N1-Diethyl-N4-(3-phenylpropyl)-1,4-benzenediamine
N1,N1-Diethyl-N4-(3-phenylpropyl)-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₉H₂₆N₂
MDL Number: MFCD03210863
CAS Number: 356532-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR