N-[4-(tert-Butyl)benzyl]-3-(2-ethoxyethoxy)aniline

Supplier: Matrix Scientific

032196-500MG
101833-216EA 306.15 USD
101833-216
N-[4-(tert-Butyl)benzyl]-3-(2-ethoxyethoxy)aniline
N-[4-(tert-Butyl)benzyl]-3-(2-ethoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MDL Number: MFCD10688202
CAS Number: 1040692-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR