N-[3-(2-Ethoxyethoxy)phenyl]-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]amine
Supplier: Matrix Scientific
032195-500MG
101833-214EA
306.15
USD
101833-214
N-[3-(2-Ethoxyethoxy)phenyl]-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]amine
N-[3-(2-Ethoxyethoxy)phenyl]-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]amine
Hazard: Irritant
Formula:
C₂₂H₃₁NO₃ |
MDL Number:
MFCD10688201 CAS Number: 1040691-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...