N-(4-Butoxybenzyl)-2-(2-ethoxyethoxy)aniline

Supplier: Matrix Scientific

032185-500MG
101833-194EA 306.15 USD
101833-194
N-(4-Butoxybenzyl)-2-(2-ethoxyethoxy)aniline
N-(4-Butoxybenzyl)-2-(2-ethoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₃
MDL Number: MFCD10688191
CAS Number: 1040691-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR