N-[2-(Phenethyloxy)benzyl]-3-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032183-500MG
101833-190EA 306.15 USD
101833-190
N-[2-(Phenethyloxy)benzyl]-3-(3-phenylpropoxy)aniline
N-[2-(Phenethyloxy)benzyl]-3-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₃₀H₃₁NO₂
MDL Number: MFCD10688189
CAS Number: 1040691-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR