N-[2-(2-Chlorophenoxy)propyl]-3-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032180-500MG
101833-184EA 306.15 USD
101833-184
N-[2-(2-Chlorophenoxy)propyl]-3-(3-phenylpropoxy)aniline
N-[2-(2-Chlorophenoxy)propyl]-3-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₄H₂₆ClNO₂
MDL Number: MFCD10688186
CAS Number: 1040690-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR