N-[2-(2-Ethoxyethoxy)benzyl]-3,4-dimethylaniline

Supplier: Matrix Scientific

032178-500MG
101833-180EA 306.15 USD
101833-180
N-[2-(2-Ethoxyethoxy)benzyl]-3,4-dimethylaniline
N-[2-(2-Ethoxyethoxy)benzyl]-3,4-dimethylaniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10688184
CAS Number: 1040690-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR