N-[2-(2-Methylphenoxy)propyl]-3-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032177-500MG
101833-178EA 306.15 USD
101833-178
N-[2-(2-Methylphenoxy)propyl]-3-(3-phenylpropoxy)aniline
N-[2-(2-Methylphenoxy)propyl]-3-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₂
MDL Number: MFCD10688183
CAS Number: 1040690-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR