N-[4-(tert-Butyl)benzyl]-3-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032176-500MG
101833-176EA 306.15 USD
101833-176
N-[4-(tert-Butyl)benzyl]-3-(3-phenylpropoxy)aniline
N-[4-(tert-Butyl)benzyl]-3-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₆H₃₁NO
MW: 373.54 g/mol
MDL Number: MFCD10688182
CAS Number: 1040690-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR