N-Benzyl-3-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032175-500MG
101833-174EA 306.15 USD
101833-174
N-Benzyl-3-(3-phenylpropoxy)aniline
N-Benzyl-3-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₂H₂₃NO
MDL Number: MFCD10688181
CAS Number: 1040690-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR