2-(Benzyloxy)-N-[2-(2-methoxyethoxy)benzyl]aniline

Supplier: Matrix Scientific

032171-500MG
101833-166EA 306.15 USD
101833-166
2-(Benzyloxy)-N-[2-(2-methoxyethoxy)benzyl]aniline
2-(Benzyloxy)-N-[2-(2-methoxyethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₃
MDL Number: MFCD10688177
CAS Number: 1040690-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR