2-(Benzyloxy)-N-[2-(4-methoxyphenoxy)ethyl]aniline

Supplier: Matrix Scientific

032170-500MG
101833-164EA 306.15 USD
101833-164
2-(Benzyloxy)-N-[2-(4-methoxyphenoxy)ethyl]aniline
2-(Benzyloxy)-N-[2-(4-methoxyphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₂₂H₂₃NO₃
MDL Number: MFCD10688176
CAS Number: 1040690-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR