2-(Benzyloxy)-N-[2-(1-naphthyl)ethyl]aniline

Supplier: Matrix Scientific

032169-500MG
101833-162EA 306.15 USD
101833-162
2-(Benzyloxy)-N-[2-(1-naphthyl)ethyl]aniline
2-(Benzyloxy)-N-[2-(1-naphthyl)ethyl]aniline

Hazard: Irritant

Formula: C₂₅H₂₃NO
MDL Number: MFCD10688175
CAS Number: 1040690-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR