N-(2-Butoxyphenyl)-N-propylamine

Supplier: Matrix Scientific

032167-500MG
101833-158EA 306.15 USD
101833-158
N-(2-Butoxyphenyl)-N-propylamine
N-(2-Butoxyphenyl)-N-propylamine

Hazard: Irritant

Formula: C₁₃H₂₁NO
MDL Number: MFCD10688173
CAS Number: 1040689-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR