2-Butoxy-N-(3,5-dichlorobenzyl)aniline

Supplier: Matrix Scientific

032165-500MG
101833-154EA 306.15 USD
101833-154
2-Butoxy-N-(3,5-dichlorobenzyl)aniline
2-Butoxy-N-(3,5-dichlorobenzyl)aniline

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO
MDL Number: MFCD10688171
CAS Number: 1040689-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR