2-Butoxy-N-[3-(2-phenoxyethoxy)benzyl]aniline

Supplier: Matrix Scientific

032163-500MG
101833-150EA 306.15 USD
101833-150
2-Butoxy-N-[3-(2-phenoxyethoxy)benzyl]aniline
2-Butoxy-N-[3-(2-phenoxyethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₃
MDL Number: MFCD10688169
CAS Number: 1040689-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR