2-Butoxy-N-(4-isobutoxybenzyl)aniline

Supplier: Matrix Scientific

032162-500MG
101833-148EA 306.15 USD
101833-148
2-Butoxy-N-(4-isobutoxybenzyl)aniline
2-Butoxy-N-(4-isobutoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MDL Number: MFCD10688168
CAS Number: 1040689-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR