N-(2-Butoxyphenyl)-N-butylamine

Supplier: Matrix Scientific

032155-500MG
101833-134EA 306.15 USD
101833-134
N-(2-Butoxyphenyl)-N-butylamine
N-(2-Butoxyphenyl)-N-butylamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10688161
CAS Number: 155593-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR