3-(Benzyloxy)-N-(3-propoxybenzyl)aniline

Supplier: Matrix Scientific

032150-500MG
101833-124EA 306.15 USD
101833-124
3-(Benzyloxy)-N-(3-propoxybenzyl)aniline
3-(Benzyloxy)-N-(3-propoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₂
MDL Number: MFCD10688156
CAS Number: 1040688-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR