3-(Benzyloxy)-N-(4-ethoxybenzyl)aniline

Supplier: Matrix Scientific

032147-500MG
101833-118EA 306.15 USD
101833-118
3-(Benzyloxy)-N-(4-ethoxybenzyl)aniline
3-(Benzyloxy)-N-(4-ethoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₂H₂₃NO₂
MDL Number: MFCD10688153
CAS Number: 1040688-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR