3-(Benzyloxy)-N-[2-(2,5-dimethylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

032144-500MG
101833-112EA 306.15 USD
101833-112
3-(Benzyloxy)-N-[2-(2,5-dimethylphenoxy)ethyl]aniline
3-(Benzyloxy)-N-[2-(2,5-dimethylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₂
MW: 347.46 g/mol
MDL Number: MFCD10688150
CAS Number: 1040688-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR