3-(Benzyloxy)-N-[2-(4-chloro-2-methylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

032143-500MG
101833-110EA 306.15 USD
101833-110
3-(Benzyloxy)-N-[2-(4-chloro-2-methylphenoxy)ethyl]aniline
3-(Benzyloxy)-N-[2-(4-chloro-2-methylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₂₂H₂₂ClNO₂
MDL Number: MFCD10688149
CAS Number: 1040688-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR